(2-Bromo-3-fluoropyridin-4-yl)boronic acid 97%

Supplier: Ambeed
Warning

A442858-1G A442858-250MG A442858-5G
77196-402EA 90 USD
77196-402 77196-404 77196-406
(2-Bromo-3-fluoropyridin-4-yl)boronic acid 97%
(2-Bromo-3-fluoropyridin-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₅H₄BBrFNO₂
MDL Number: MFCD18434408
CAS Number: 2096338-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR