tert-Butyl 4-bromo-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A442352-100MG
A442352-1G
A442352-250MG
A442352-25MG
A442352-50MG
77196-392EA
289.43
USD
77196-392
77196-394
77196-396
77196-398
77196-400
tert-Butyl 4-bromo-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate 97%
tert-Butyl 4-bromo-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate
Precaución: for Research Use Only
Formula:
C₁₇H₂₁BrN₂O₃ MW: 381.26 g/mol |
MDL Number:
MFCD27995898 CAS Number: 1707580-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...