5-Bromo-N-quinolin-8-ylthiophene-2-sulfonamide 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-N-(quinolin-8-yl)thiophene-2-sulphonamide

A442030-1G A442030-250MG
77196-384EA 144.2 USD
77196-384 77196-386
5-Bromo-N-quinolin-8-ylthiophene-2-sulfonamide 97%
5-Bromo-N-quinolin-8-ylthiophene-2-sulfonamide

Precaución: for Research Use Only

Formula: C₁₃H₉BrN₂O₂S₂
MW: 369.27 g/mol
MDL Number: MFCD06290688
CAS Number: 620103-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR