2-Chloro-4-isopropoxy-5-nitrotoluene 97%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-4-methyl-2-nitrophenyl prop-2-yl ether, 4-Chloro-5-methyl-2-[(prop-2-y)loxy]nitrobenzene, 1-Chloro-5-isopropoxy-2-methyl-4-nitrobenzene

A441695-250MG A441695-1G
77168-798EA 7.21 USD
77168-798 77684-216
2-Chloro-4-isopropoxy-5-nitrotoluene 97%
2-Chloro-4-isopropoxy-5-nitrotoluene

Precaución: For research use only.

Formula: C₁₀H₁₂ClNO₃
MW: 229.66 g/mol
MDL Number: MFCD11499001
CAS Number: 1032903-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR