2-Amino-3-iodobenzonitrile 97%

Supplier: Ambeed
Warning

A441583-1G A441583-5G
77196-380EA 19 USD
77196-380 77196-382
2-Amino-3-iodobenzonitrile 97%
2-Amino-3-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅IN₂
MDL Number: MFCD13689143
CAS Number: 114344-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR