4-Methyl-3-nitrophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-Methyl-3-nitrophenyl)boronic acid, 4-Methyl-3-nitrobenzeneboronic acid

A441356-1G A441356-5G
77196-378EA 6 USD
77196-378 77746-901
4-Methyl-3-nitrophenylboronic acid 98%
4-Methyl-3-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BNO₄
MDL Number: MFCD00191550
CAS Number: 80500-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR