1-Chloro-4-(chloromethyl)-2-methylbenzene 95%

Supplier: Ambeed
Danger

A441238-100MG A441238-1G A441238-250MG
77168-792EA 40 USD
77168-792 77168-794 77168-796
1-Chloro-4-(chloromethyl)-2-methylbenzene 95%
1-Chloro-4-(chloromethyl)-2-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈Cl₂
MDL Number: MFCD11846567
CAS Number: 92304-76-2
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR