[1,1'-Bi(cyclopropane)]-1-carboxylic acid 97%

Supplier: Ambeed
Warning

A441030-100MG A441030-1G A441030-250MG A441030-5G
77168-784EA 107.12 USD
77168-784 77168-786 77168-788 77168-790
[1,1'-Bi(cyclopropane)]-1-carboxylic acid 97%
[1,1'-Bi(cyclopropane)]-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₀O₂
MW: 126.15 g/mol
MDL Number: MFCD08704251
CAS Number: 60629-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR