tert-Butyl (5-amino-2-methylphenyl)carbamate 97%

Supplier: Ambeed
Warning

A440692-1G A440692-250MG A440692-5G
77168-766EA 27 USD
77168-766 77168-768 77168-770
tert-Butyl (5-amino-2-methylphenyl)carbamate 97%
tert-Butyl (5-amino-2-methylphenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₂
MDL Number: MFCD04114553
CAS Number: 329763-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR