(6-(3-Fluorophenoxy)pyridin-3-yl)boronic acid 97%

Supplier: Ambeed
Warning

A440317-1G A440317-250MG A440317-5G
77196-354EA 164 USD
77196-354 77196-356 77196-358
(6-(3-Fluorophenoxy)pyridin-3-yl)boronic acid 97%
(6-(3-Fluorophenoxy)pyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₉BFNO₃
MDL Number: MFCD22375012
CAS Number: 2051588-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR