2-Hydroxy-7-methoxyquinoline-3-carboxaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-7-methoxyquinoline-3-carbaldehyde, 3-Formyl-2-hydroxy-7-methoxyquinoline, 3-Formyl-2-hydroxy-7-methoxy-1-azanaphthalene

A440191-100MG A440191-1G A440191-250MG A440191-5G
77168-758EA 73 USD
77168-758 77168-760 77168-762 77168-764
2-Hydroxy-7-methoxyquinoline-3-carboxaldehyde 95%
2-Hydroxy-7-methoxyquinoline-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₉NO₃
MW: 203.194 g/mol
MDL Number: MFCD02986395
CAS Number: 101382-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR