4-Hydroxysophthalic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxyisophthalic acid

A439933-100G A439933-10G A439933-1G A439933-25G A439933-5G
77168-748EA 282.99 USD
77168-748 77168-750 77168-752 77168-754 77168-756
4-Hydroxysophthalic acid 98%
4-Hydroxysophthalic acid

Precaución: for Research Use Only

Formula: C₈H₆O₅
MDL Number: MFCD00010391
CAS Number: 636-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR