Methyl 2-cyano-6-fluorobenzoate 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A439641-5G A439641-1G
77168-746EA 46 USD
77168-746 77684-211
Methyl 2-cyano-6-fluorobenzoate 98%, Ambeed.Inc
Methyl 2-cyano-6-fluorobenzoate

Precaución: For research use only.

Formula: C₉H₆FNO₂
MW: 179.15 g/mol
MDL Number: MFCD12911464
CAS Number: 58332-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR