Palladium(II) hexafluoroacetylacetonate 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A439163-100MG A439163-250MG A439163-1G
77168-742EA 29 USD
77168-742 77168-744 77684-209
Palladium(II) hexafluoroacetylacetonate 98%, Ambeed.Inc
Palladium(II) hexafluoroacetylacetonate

Precaución: For research use only.

Formula: C₁₀H₂F₁₂O₄Pd
MDL Number: MFCD00015709
CAS Number: 64916-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR