[2-Chloro-6-(trifluoromethyl)pyridin-4-yl]boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-Chloro-6-(trifluoromethyl)pyridin-4-yl)boronic acid, 2-Chloro-6-(trifluoromethyl)pyridine-4-boronic acid

A438501-100MG A438501-1G A438501-250MG
77196-342EA 39 USD
77196-342 77196-344 77196-346
[2-Chloro-6-(trifluoromethyl)pyridin-4-yl]boronic acid 98%
[2-Chloro-6-(trifluoromethyl)pyridin-4-yl]boronic acid

Precaución: for Research Use Only

Formula: C₆H₄BClF₃NO₂
MW: 225.361 g/mol
MDL Number: MFCD13188845
CAS Number: 1446486-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR