11H-Benzo[a]carbazole 98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Benzocarbazole

A438095-1G A438095-5G
77196-336EA 28.84 USD
77196-336 77196-338
11H-Benzo[a]carbazole 98%
11H-Benzo[a]carbazole

Precaución: for Research Use Only

Formula: C₁₆H₁₁N
MW: 217.27 g/mol
MDL Number: MFCD00215939
CAS Number: 239-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR