(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)methanamine 95%

Supplier: Ambeed
Danger

A437338-1G A437338-250MG
77168-736EA 96 USD
77168-736 77168-738
(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)methanamine 95%
(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)methanamine

Precaución: for Research Use Only

Formula: C₈H₇F₂NO₂
MDL Number: MFCD06212945
CAS Number: 135132-35-3
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR