4,5-Dibromo-3-methylthiophene-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A436096-1G A436096-250MG A436096-5G
77168-730EA 18 USD
77168-730 77168-732 77168-734
4,5-Dibromo-3-methylthiophene-2-carboxylic acid 95%
4,5-Dibromo-3-methylthiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₄Br₂O₂S
MDL Number: MFCD27930042
CAS Number: 854626-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR