tert-Butyl (1-(2-hydroxyethyl)cyclopropyl)carbamate 95%

Supplier: Ambeed
Warning

A434804-100MG A434804-1G A434804-250MG A434804-5G
77168-714EA 28 USD
77168-714 77168-716 77168-718 77892-905
tert-Butyl (1-(2-hydroxyethyl)cyclopropyl)carbamate 95%
tert-Butyl (1-(2-hydroxyethyl)cyclopropyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MDL Number: MFCD24444732
CAS Number: 753023-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR