1-(tert-Butyl)-3-methyl-1H-pyrazol-5-ylamine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(tert-Butyl)-3-methyl-1H-pyrazol-5-amine
A434721-1G
A434721-250MG
A434721-25G
A434721-5G
77196-320EA
20.6
USD
77196-320
77196-322
77196-324
77196-326
1-(tert-Butyl)-3-methyl-1H-pyrazol-5-ylamine 98%
1-(tert-Butyl)-3-methyl-1H-pyrazol-5-ylamine
Precaución: for Research Use Only
Formula:
C₈H₁₅N₃ MW: 153.23 g/mol |
MDL Number:
MFCD00122731 CAS Number: 141459-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...