Diethyl-2-chloro-3-oxosuccinate 90%

Supplier: Ambeed
Danger

A433805-1G A433805-5G
77168-704EA 9 USD
77168-704 77168-706
Diethyl-2-chloro-3-oxosuccinate 90%
Diethyl-2-chloro-3-oxosuccinate

Precaución: for Research Use Only

Formula: C₈H₁₁ClO₅
MDL Number: MFCD04114359
CAS Number: 34034-87-2
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR