Ethyl 4-chloro-7-(phenylsulfonyl)-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylate 95%

Supplier: Ambeed
Warning

A433392-100MG A433392-1G A433392-250MG
77168-698EA 27.81 USD
77168-698 77168-700 77168-702
Ethyl 4-chloro-7-(phenylsulfonyl)-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylate 95%
Ethyl 4-chloro-7-(phenylsulfonyl)-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₂ClN₃O₄S
MW: 365.79 g/mol
MDL Number: MFCD19703862
CAS Number: 1987286-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR