2,3-Dihydro-1H-pyrrolo[3,4-c]pyridine hydrochloride 97%

Supplier: Ambeed
Warning

A433069-100MG A433069-250MG A433069-1G
77168-684EA 49.44 USD
77168-684 77168-686 77684-204
2,3-Dihydro-1H-pyrrolo[3,4-c]pyridine hydrochloride 97%
2,3-Dihydro-1H-pyrrolo[3,4-c]pyridine hydrochloride

Precaución: For research use only.

Formula: C₇H₉ClN₂
MDL Number: MFCD08273906
CAS Number: 651558-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR