N-Succinimidyl [(tert-Butoxycarbonyl)aminooxy]acetate 98%

Supplier: Ambeed
Warning

Synonyms: N-Succinimidyl (Boc-aminooxy)acetate, 2,5-Dioxopyrrolidin-1-yl 2-(((tert-butoxycarbonyl)amino)oxy)acetate, (Boc-aminooxy)acetic acid N-Succinimidyl ester, [(tert-Butoxycarbonyl)aminooxy]acetic acid N-Succinimidyl ester, Boc-NH-O-C1-NHS ester

A433010-1G A433010-250MG
77196-314EA 61.99 USD
77196-314 77746-880
N-Succinimidyl [(tert-Butoxycarbonyl)aminooxy]acetate 98%
N-Succinimidyl [(tert-Butoxycarbonyl)aminooxy]acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂O₇
MDL Number: MFCD18382162
CAS Number: 80366-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR