1-Bromo-2,4-bis(trifluoromethyl)benzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,4-Bis(trifluoromethyl)-1-bromobenzene
A432560-10G
A432560-1G
A432560-25G
A432560-5G
77196-308EA
41.2
USD
77196-308
77196-310
77196-312
77746-879
1-Bromo-2,4-bis(trifluoromethyl)benzene 98%
1-Bromo-2,4-bis(trifluoromethyl)benzene
Precaución: for Research Use Only
Formula:
C₈H₃BrF₆ MW: 293.01 g/mol |
MDL Number:
MFCD00074904 CAS Number: 327-75-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...