Methyl-1H-indazole-5-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A432555-100G
A432555-10G
A432555-1G
A432555-25G
A432555-500G
A432555-5G
77168-658EA
199
USD
77168-658
77168-660
77168-662
77168-664
77168-666
77168-668
Methyl-1H-indazole-5-carboxylate 98%
Methyl 1H-indazole-5-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₈N₂O₂ |
MDL Number:
MFCD07371609 CAS Number: 473416-12-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...