N1-(3,4-Dimethoxybenzyl)-N3,N3-dimethylpropane-1,3-diamine 97%

Supplier: Ambeed
Warning

A432009-100MG A432009-1G A432009-250MG A432009-5G
77196-300EA 15 USD
77196-300 77196-302 77196-304 77196-306
N1-(3,4-Dimethoxybenzyl)-N3,N3-dimethylpropane-1,3-diamine 97%
N1-(3,4-Dimethoxybenzyl)-N3,N3-dimethylpropane-1,3-diamine

Precaución: for Research Use Only

Formula: C₁₄H₂₄N₂O₂
MW: 252.35 g/mol
MDL Number: MFCD07406424
CAS Number: 46907-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR