4-Ethyl-N-(3-((4-methylpiperazin-1-yl)methyl)-5-(trifluoromethyl)phenyl)-3-(pyrazolo[1,5-a]pyrimidin-6-ylethynyl)benzamide 97%

Supplier: Ambeed
Warning

Synonyms: 7rh

A431837-1MG A431837-25MG A431837-5MG A431837-10MG A431837-50MG
77196-294EA 29.87 USD
77196-294 77196-296 77196-298 77746-871 77746-872
4-Ethyl-N-(3-((4-methylpiperazin-1-yl)methyl)-5-(trifluoromethyl)phenyl)-3-(pyrazolo[1,5-a]pyrimidin-6-ylethynyl)benzamide 97%
4-Ethyl-N-(3-((4-methylpiperazin-1-yl)methyl)-5-(trifluoromethyl)phenyl)-3-(pyrazolo[1,5-a]pyrimidin-6-ylethynyl)benzamide

Precaución: for Research Use Only

Formula: C₃₀H₂₉F₃N₆O
MW: 546.59 g/mol
MDL Number: MFCD30185014
CAS Number: 1429617-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR