5-Bromo-1H-indole-2-carbaldehyde 97%

Supplier: Ambeed
Warning

A430801-1G A430801-5G A430801-100MG A430801-250MG
77168-648EA 211.15 USD
77168-648 77168-650 77684-200 77684-201
5-Bromo-1H-indole-2-carbaldehyde 97%
5-Bromo-1H-indole-2-carbaldehyde

Precaución: For research use only.

Formula: C₉H₆BrNO
MDL Number: MFCD06738773
CAS Number: 53590-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR