4-Phenyloxazolidin-2-one 98%

Supplier: Ambeed
Warning

A430328-100G A430328-1G A430328-25G A430328-5G
77168-628EA 290.46 USD
77168-628 77168-630 77168-632 77168-634
4-Phenyloxazolidin-2-one 98%
4-Phenyloxazolidin-2-one

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.17 g/mol
MDL Number: MFCD00601044
CAS Number: 7480-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR