2-(4-Bromophenyl)piperidine 97%

Supplier: Ambeed
Warning

A429975-100MG A429975-1G A429975-250MG A429975-5G
77168-620EA 35 USD
77168-620 77168-622 77168-624 77892-894
2-(4-Bromophenyl)piperidine 97%
2-(4-Bromophenyl)piperidine

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrN
MDL Number: MFCD02663580
CAS Number: 383128-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR