Benzyl 4-(sulfamoylmethyl)piperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A429612-100MG A429612-1G A429612-250MG
77168-614EA 40.17 USD
77168-614 77168-616 77168-618
Benzyl 4-(sulfamoylmethyl)piperidine-1-carboxylate 95%
Benzyl 4-(sulfamoylmethyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₄S
MW: 312.385 g/mol
MDL Number: MFCD27501042
CAS Number: 1585225-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR