Benzo[d][1,2,3]thiadiazole 97%

Supplier: Ambeed
Warning

A429526-100MG A429526-1G A429526-250MG
77196-282EA 16 USD
77196-282 77196-284 77196-286
Benzo[d][1,2,3]thiadiazole 97%
Benzo[d][1,2,3]thiadiazole

Precaución: for Research Use Only

Formula: C₆H₄N₂S
MDL Number: MFCD00047265
CAS Number: 273-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR