3-Iodo-2-nitroaniline 97%

Supplier: Ambeed
Danger

A429110-100MG A429110-250MG
77196-272EA 12 USD
77196-272 77196-274
3-Iodo-2-nitroaniline 97%
3-Iodo-2-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₅IN₂O₂
MDL Number: MFCD18397930
CAS Number: 721925-17-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR