3,6-Dimethyl-2,5-piperazinedione 98%

Supplier: Ambeed
Warning

Synonyms: Alanine anhydride, 3,6-Dimethylpiperazine-2,5-dione, 3,6-Dimethyl-2,5-piperazinedione

A428887-1G A428887-25G A428887-5G
77168-610EA 16.48 USD
77168-610 77684-196 77684-197
3,6-Dimethyl-2,5-piperazinedione 98%
3,6-Dimethyl-2,5-piperazinedione

Precaución: For research use only.

Formula: C₆H₁₀N₂O₂
MW: 142.16 g/mol
MDL Number: MFCD00023153
CAS Number: 5625-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR