(5-(Trifluoromethyl)thiophen-2-yl)boronic acid 95%

Supplier: Ambeed
Warning

A428773-1G A428773-250MG A428773-50MG A428773-100MG
77168-604EA 429.51 USD
77168-604 77168-606 77168-608 77684-195
(5-(Trifluoromethyl)thiophen-2-yl)boronic acid 95%
(5-(Trifluoromethyl)thiophen-2-yl)boronic acid

Precaución: For research use only.

Formula: C₅H₄BF₃O₂S
MW: 195.95 g/mol
MDL Number: MFCD11042442
CAS Number: 958451-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR