5-Hexylthiophene-2-carbaldehyde 95%

Supplier: Ambeed
Danger

Synonyms: 5-Hexylthiophene-2-carboxaldehyde

A428706-1G A428706-25G
77196-268EA 15.45 USD
77196-268 77196-270
5-Hexylthiophene-2-carbaldehyde 95%
5-Hexylthiophene-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₆OS
MW: 196.30 g/mol
MDL Number: MFCD11044868
CAS Number: 100943-46-2
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR