(1R)-1-Pyridin-4-ylethanamine 95%

Supplier: Ambeed
Danger

Synonyms: (R)-1-(4-Pyridyl)ethylamine

A427540-100MG A427540-1G A427540-5G
77168-602EA 32.96 USD
77168-602 77684-191 77684-192
(1R)-1-Pyridin-4-ylethanamine 95%
(1R)-1-Pyridin-4-ylethanamine

Precaución: For research use only.

Formula: C₇H₁₀N₂
MW: 122.168 g/mol
MDL Number: MFCD09256818
CAS Number: 45682-36-8
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR