N-Formylsaccharin 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A427512-10G A427512-1G A427512-5G A427512-25G A427512-100G A427512-250MG
77168-596EA 57 USD
77168-596 77168-598 77168-600 77684-190 77892-886 77892-887
N-Formylsaccharin 95%, Ambeed.Inc
N-Formylsaccharin

Precaución: For research use only.

Formula: C₈H₅NO₄S
MDL Number: MFCD23380208
CAS Number: 50978-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR