But-3-en-1-amine hydrochloride 95%

Supplier: Ambeed
Danger

A426953-100MG A426953-10G A426953-1G A426953-250MG A426953-25G A426953-5G
77196-256EA 5.15 USD
77196-256 77196-258 77196-260 77196-262 77196-264 77196-266
But-3-en-1-amine hydrochloride 95%
But-3-en-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₉N·ClH
MW: 107.582 g/mol
MDL Number: MFCD05663670
CAS Number: 17875-18-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR