Benzo[b]thiophene-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A426586-100MG A426586-250MG A426586-50MG A426586-1G
77196-248EA 32.96 USD
77196-248 77196-250 77196-252 77746-858
Benzo[b]thiophene-5-carboxylic acid 97%
Benzo[b]thiophene-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆O₂S
MW: 178.2077 g/mol
MDL Number: MFCD04974042
CAS Number: 2060-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR