Methyl 3-iodothiophene-2-carboxylate 95%

Supplier: Ambeed
Warning

A425374-1G A425374-250MG A425374-5G A425374-25G
77168-584EA 41.2 USD
77168-584 77168-586 77168-588 77613-314
Methyl 3-iodothiophene-2-carboxylate 95%
Methyl 3-iodothiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₅IO₂S
MW: 268.07 g/mol
MDL Number: MFCD03407485
CAS Number: 62353-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR