Gilteritinib 97%

Supplier: Ambeed
Warning

Synonyms: 6-Ethyl-3-((3-methoxy-4-(4-(4-methylpiperazin-1-yl)piperidin-1-yl)phenyl)amino)-5-((tetrahydro-2H-pyran-4-yl)amino)pyrazine-2-carboxamide, ASP 2215

A425350-10MG A425350-2MG A425350-1MG A425350-250MG A425350-50MG
77168-580EA 46.35 USD
77168-580 77168-582 77684-185 77684-186 77684-187
Gilteritinib 97%
Gilteritinib

Precaución: For research use only.

Formula: C₂₉H₄₄N₈O₃
MW: 552.71 g/mol
MDL Number: MFCD28144685
CAS Number: 1254053-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR