Methyl 3-cyanoisonicotinate 95%

Supplier: Ambeed
Warning

A424810-100MG A424810-1G A424810-250MG
77168-574EA 38.11 USD
77168-574 77168-576 77168-578
Methyl 3-cyanoisonicotinate 95%
Methyl 3-cyanoisonicotinate

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MW: 162.14 g/mol
MDL Number: MFCD00128866
CAS Number: 36106-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR