Rucaparib phosphate ≥99%

Supplier: Ambeed
Warning

Synonyms: 8-Fluoro-2-(4-((methylamino)methyl)phenyl)-4,5-dihydro-1H-azepino[5,4,3-cd]indol-6(3H)-one phosphate

A424427-10MG A424427-1MG A424427-25MG A424427-50MG A424427-5MG
77168-564EA 90.64 USD
77168-564 77168-566 77168-568 77168-570 77168-572
Rucaparib phosphate ≥99%
Rucaparib phosphate

Precaución: for Research Use Only

Formula: C₁₉H₂₁FN₃O₅P
MW: 421.3593 g/mol
MDL Number: MFCD17010269
CAS Number: 459868-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR