1-(Phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine-6-carbonitrile 95%

Supplier: Ambeed
Warning

A424014-100MG A424014-1G A424014-250MG
77196-242EA 57.68 USD
77196-242 77196-244 77196-246
1-(Phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine-6-carbonitrile 95%
1-(Phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine-6-carbonitrile

Precaución: for Research Use Only

Formula: C₁₄H₉N₃O₂S
MW: 283.31 g/mol
MDL Number: MFCD16875751
CAS Number: 1227270-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR