(Quinolin-2-ylmethyl)amine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Aminomethyl)quinoline hydrochloride, Quinolin-2-ylmethanamine hydrochloride

A422152-100MG A422152-1G A422152-250MG
77196-236EA 66.95 USD
77196-236 77196-238 77196-240
(Quinolin-2-ylmethyl)amine hydrochloride 97%
(Quinolin-2-ylmethyl)amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClN₂
MW: 194.66 g/mol
MDL Number: MFCD09966142
CAS Number: 861036-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR