2,5-Dioxopyrrolidin-1-yl 4-(3-(trifluoromethyl)-3H-diazirin-3-yl)benzoate 97%

Supplier: Ambeed
Danger

Synonyms: 4-[3-(Trifluoromethyl)diazirin-3-yl] benzoic acid N-hydroxysuccinimide ester

A421872-100MG A421872-250MG
77196-232EA 98.88 USD
77196-232 77196-234
2,5-Dioxopyrrolidin-1-yl 4-(3-(trifluoromethyl)-3H-diazirin-3-yl)benzoate 97%
2,5-Dioxopyrrolidin-1-yl 4-(3-(trifluoromethyl)-3H-diazirin-3-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₃H₈F₃N₃O₄
MW: 327.21 g/mol
MDL Number: MFCD01075064
CAS Number: 87736-89-8
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR