(2S,3R,4S,5R,6R)-6-(((3,4,5-Trihydroxybenzoyl)oxy)methyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetrakis(3,4,5-trihydroxybenzoate) 98%

Supplier: Ambeed
Warning

Synonyms: 1,2,3,4,6-O-pentagalloylglucose

A421340-10MG A421340-1MG A421340-250MG A421340-50MG A421340-5MG
77196-222EA 54.59 USD
77196-222 77196-224 77196-226 77196-228 77196-230
(2S,3R,4S,5R,6R)-6-(((3,4,5-Trihydroxybenzoyl)oxy)methyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetrakis(3,4,5-trihydroxybenzoate) 98%
(2S,3R,4S,5R,6R)-6-(((3,4,5-Trihydroxybenzoyl)oxy)methyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetrakis(3,4,5-trihydroxybenzoate)

Precaución: for Research Use Only

Formula: C₄₁H₃₂O₂₆
MW: 940.67 g/mol
MDL Number: MFCD03788860
CAS Number: 14937-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR