5,11-Dihydroindolo[3,2-b]carbazole 97%

Supplier: Ambeed
Warning

Synonyms: Indolo[3,2-b]carbazole

A420590-100G A420590-10G A420590-1G A420590-25G A420590-5G
77168-536EA 1347.24 USD
77168-536 77168-538 77168-540 77168-542 77168-544
5,11-Dihydroindolo[3,2-b]carbazole 97%
5,11-Dihydroindolo[3,2-b]carbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₂N₂
MW: 256.31 g/mol
MDL Number: MFCD09879258
CAS Number: 6336-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR